SY3H3D67CF8B Synthonix Corporation

9,9-Dimethyl-2-nitro-9H-fluorene

  • MDL number MFCD17170053
  • Cas Number : 605644-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58193.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58193.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart