SY3H3D67CF8C Synthonix Corporation

9,9-Dimethyl-9H-fluorene

  • MDL number MFCD00114670
  • Cas Number : 4569-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58214.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/58214.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart