SY3H3D67CF8E Synthonix Corporation

(E)-1,3-Diphenylprop-2-en-1-ol

  • Cas Number : 62668-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58226.html

Safety Information

MSDS https://synthonix.com/58226.html
Availability details for
Quantity
  • Availability
  • Add to Cart