SY3H3D67CF96 Synthonix Corporation

1,3-Dimethyl-1,3-diphenylurea

  • Cas Number : 611-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58268.html

Safety Information

MSDS https://synthonix.com/58268.html
Availability details for
Quantity
  • Availability
  • Add to Cart