SY3H3D67CF9D Synthonix Corporation

N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-ethylacetamide

  • MDL number MFCD04117958
  • Cas Number : 96605-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Slightly pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58366.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58366.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart