SY3H3D67CFA0 Synthonix Corporation

Di-tert-butyl 5-methyl-2,4-dioxopyrimidine-1,3(2H,4H)-dicarboxylate

  • MDL number MFCD30180247
  • Cas Number : 917081-58-4
  • Molecular Weight : 326.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name di-tert-butyl 5-methyl-2,4-dioxopyrimidine-1,3(2H,4H)-dicarboxylate
Inchi Code 1S/C15H22N2O6/c1-9-8-16(12(20)22-14(2,3)4)11(19)17(10(9)18)13(21)23-15(5,6)7/h8H,1-7H3
Inchi Key MBNPMTPOCIYNPX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58433.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/58433.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart