SY3H3D67CFA7 Synthonix Corporation

1,4-Di-Boc-piperazine-2-carboxylic acid

  • MDL number MFCD02179115
  • Cas Number : 181955-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58494.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58494.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart