SY3H3D67CFA8 Synthonix Corporation

Dicyclohexylamine (S)-6-(((allyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate

  • MDL number MFCD00273410
  • Cas Number : 110637-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58496.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58496.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart