SY3H3D67CFBA Synthonix Corporation

4-(3,4-Dichlorophenyl)-4-phenylbutanoic acid

  • MDL number MFCD09833046
  • Cas Number : 79560-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58638.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58638.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart