SY3H3D67CFBE Synthonix Corporation

5,6-Diphenylmorpholin-2-one

  • Cas Number : 19180-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58653.html

Safety Information

MSDS https://synthonix.com/58653.html
Availability details for
Quantity
  • Availability
  • Add to Cart