SY3H3D67CFCC Synthonix Corporation

Diisobutyl terephthalate

  • MDL number MFCD01682719
  • Cas Number : 18699-48-4
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diisobutyl terephthalate
Inchi Code 1S/C16H22O4/c1-11(2)9-19-15(17)13-5-7-14(8-6-13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3
Inchi Key LQKWPGAPADIOSS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58793.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/58793.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart