SY3H3D67CFE4 Synthonix Corporation

4-((2-(Dimethylamino)ethoxy)(phenyl)methyl)aniline

  • MDL number MFCD22373419
  • Cas Number : 101602-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59023.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59023.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart