SY3H3D67CFEC Synthonix Corporation

6,12-Dibromochrysene

  • Cas Number : 131222-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59114.html

Safety Information

MSDS https://synthonix.com/59114.html
Availability details for
Quantity
  • Availability
  • Add to Cart