SY3H3D67CFED Synthonix Corporation

2,7-Dibromotriphenylene

  • Cas Number : 888041-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59115.html

Safety Information

MSDS https://synthonix.com/59115.html
Availability details for
Quantity
  • Availability
  • Add to Cart