SY3H3D67CFF7 Synthonix Corporation

11,12-Dihydroindolo[2,3-a]carbazole

  • MDL number MFCD07371389
  • Cas Number : 60511-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59138.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59138.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart