SY3H3D67CFFC Synthonix Corporation

2,7-Dibromo-9-hexyl-9H-carbazole

  • MDL number MFCD29089359
  • Cas Number : 654676-12-7
  • Molecular Weight : 409.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,7-dibromo-9-hexyl-9H-carbazole
Inchi Code 1S/C18H19Br2N/c1-2-3-4-5-10-21-17-11-13(19)6-8-15(17)16-9-7-14(20)12-18(16)21/h6-9,11-12H,2-5,10H2,1H3
Inchi Key YSUUFISKPJBPOH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/59217.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/59217.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart