SY3H3D67D001 Synthonix Corporation

(R)-2,5-Dioxopyrrolidin-1-yl 2-(((benzyloxy)carbonyl)amino)-4-methylpentanoate

  • MDL number MFCD00153337
  • Cas Number : 65581-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59257.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59257.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart