SY3H3D67D13D Synthonix Corporation

(4-Dodecylphenyl)boronic acid

  • Cas Number : 206763-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59329.html

Safety Information

MSDS https://synthonix.com/59329.html
Availability details for
Quantity
  • Availability
  • Add to Cart