SY3H3D67D145 Synthonix Corporation

1-(2,7-Dibromospiro[fluorene-9,4'-piperidin]-1'-yl)ethanone

  • MDL number MFCD28144724
  • Cas Number : 1616114-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59382.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59382.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart