SY3H3D67D14C Synthonix Corporation

Dodecyl 4,6-dibromothieno[3,4-b]thiophene-2-carboxylate

  • MDL number MFCD16619393
  • Cas Number : 1098102-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59439.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59439.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart