SY3H3D67D14D Synthonix Corporation

2-(Dipropylamino)-2-oxoethyl 3-(((benzyloxy)carbonyl)amino)propanoate

  • MDL number MFCD30490712
  • Cas Number : 1366067-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid or Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59446.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59446.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart