SY3H3D67D14F Synthonix Corporation

Dicyclohexyl(o-tolyl)phosphine

  • Cas Number : 173593-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59453.html

Safety Information

MSDS https://synthonix.com/59453.html
Availability details for
Quantity
  • Availability
  • Add to Cart