SY3H3D67D158 Synthonix Corporation

3-(Dimethylamino)-2-(1-(phenylsulfonyl)-1H-indole-2-carbonyl)acrylonitrile

  • MDL number MFCD30345534
  • Cas Number : 1265231-91-1
  • Molecular Weight : 379.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59502.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59502.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart