SY3H3D67D15C Synthonix Corporation

(S)-4-(4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl)-3-(hydroxymethyl)benzonitrile

  • MDL number MFCD16661208
  • Cas Number : 488787-59-3
  • Molecular Weight : 342.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59555.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59555.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart