SY3H3D67D168 Synthonix Corporation

4-(Dibenzylamino)cyclohexanone hydrochloride

  • MDL number MFCD22421636
  • Cas Number : 1416352-02-7
  • Molecular Weight : 329.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(dibenzylamino)cyclohexan-1-one hydrochloride
Inchi Code 1S/C20H23NO.ClH/c22-20-13-11-19(12-14-20)21(15-17-7-3-1-4-8-17)16-18-9-5-2-6-10-18;/h1-10,19H,11-16H2;1H
Inchi Key TUTOCKYAXGAJTC-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/59615.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/59615.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart