SY3H3D67D16A Synthonix Corporation

Diethyl 2-vinylcyclopropane-1,1-dicarboxylate

  • Cas Number : 7686-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54971.html

Safety Information

MSDS https://synthonix.com/54971.html
Availability details for
Quantity
  • Availability
  • Add to Cart