SY3H3D67D185 Synthonix Corporation

(2R,3S)-Ethyl 2-amino-3-hydroxy-3-(4-(methylsulfonyl)phenyl)propanoate

  • MDL number MFCD08274466
  • Cas Number : 36983-12-7
  • Molecular Weight : 287.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (2R,3S)-2-amino-3-hydroxy-3-[4-(methylsulfonyl)phenyl]propanoate
Inchi Code 1S/C12H17NO5S/c1-3-18-12(15)10(13)11(14)8-4-6-9(7-5-8)19(2,16)17/h4-7,10-11,14H,3,13H2,1-2H3/t10-,11+/m1/s1
Inchi Key CEEHCOWSYFANRT-MNOVXSKESA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39680.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39680.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart