SY3H3D67D189 Synthonix Corporation

Ethyl phosphorodichloridate

  • MDL number No MDL found
  • Cas Number : 1498-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39780.html

Safety Information

MSDS https://synthonix.com/39780.html
Availability details for
Quantity
  • Availability
  • Add to Cart