SY3H3D67D18D Synthonix Corporation

1-Ethyl-1,2,4-triazole

  • MDL number MFCD07367971
  • Cas Number : 16778-70-4
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethyl-1H-1,2,4-triazole
Inchi Code 1S/C4H7N3/c1-2-7-4-5-3-6-7/h3-4H,2H2,1H3
Inchi Key FEOIYPLRWRCSMS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form White to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/40218.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40218.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart