SY3H3D67D1A1 Synthonix Corporation

5-Ethylisoxazol-3-amine

  • Cas Number : 19754-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41190.html

Safety Information

MSDS https://synthonix.com/41190.html
Availability details for
Quantity
  • Availability
  • Add to Cart