SY3H3D67D1AD Synthonix Corporation

1-ethyl-3-methylimidazolium iodide

  • MDL number No MDL found
  • Cas Number : 35935-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41405.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41405.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart