SY3H3D67D1B9 Synthonix Corporation

Ethyl 2-(2-chloroethoxy)acetate

  • MDL number MFCD08458712
  • Cas Number : 17229-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to Pale-yellow or Pale-green Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41531.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41531.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart