SY3H3D67D1BB Synthonix Corporation

1-Ethylpyrrolidin-3-ol

  • MDL number MFCD00003179
  • Cas Number : 30727-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow red - Red-brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41574.html

Safety Information

signalword Warning
pcodes P210
MSDS https://synthonix.com/41574.html
pictograms GHS07
hstatements H227, H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart