SY3H3D67D1CC Synthonix Corporation

Ethyl 5-chloro-1H-pyrazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 1393667-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43043.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43043.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart