SY3H3D67D1D1 Synthonix Corporation

Ethyl isoxazole-5-carboxylate

  • Cas Number : 173850-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43160.html

Safety Information

MSDS https://synthonix.com/43160.html
Availability details for
Quantity
  • Availability
  • Add to Cart