SY3H3D67D1D2 Synthonix Corporation

4-Ethylthiazole-2-carbaldehyde

  • MDL number MFCD11520452
  • Cas Number : 211943-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Liquid to Sticky Oil to Semi-Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43175.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43175.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart