SY3H3D67D225 Synthonix Corporation

1-Ethylpyrazole-3-carboxylic Acid

  • MDL number MFCD02090832
  • Cas Number : 400755-44-4
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43220.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43220.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart