SY3H3D67D22A Synthonix Corporation

Ethyl 3-methyl-1,2,4-oxadiazole-5-carboxylate

  • MDL number MFCD08275042
  • Cas Number : 40019-21-4
  • Molecular Weight : 156.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Sticky Oil to Solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43247.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43247.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart