SY3H3D67D248 Synthonix Corporation

2-Ethoxy-6-methylpyrazine

  • MDL number MFCD07787591
  • Cas Number : 53163-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43628.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43628.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart