SY3H3D67D257 Synthonix Corporation

Ethyl 2-amino-4-methyloxazole-5-carboxylate

  • MDL number MFCD09028430
  • Cas Number : 79221-15-1
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43677.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43677.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart