SY3H3D67D259 Synthonix Corporation

Ethyl 1-methyl-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 103626-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43681.html

Safety Information

MSDS https://synthonix.com/43681.html
Availability details for
Quantity
  • Availability
  • Add to Cart