SY3H3D67D25F Synthonix Corporation

Ethyl 2,2-difluoropent-4-enoate

  • Cas Number : 110482-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43722.html

Safety Information

MSDS https://synthonix.com/43722.html
Availability details for
Quantity
  • Availability
  • Add to Cart