SY3H3D67D267 Synthonix Corporation

1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole

  • MDL number MFCD22123601
  • Cas Number : 575452-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43750.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43750.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart