SY3H3D67D278 Synthonix Corporation

Ethyl morpholine-2-carboxylate

  • Cas Number : 135782-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43856.html

Safety Information

MSDS https://synthonix.com/43856.html
Availability details for
Quantity
  • Availability
  • Add to Cart