SY3H3D67D28B Synthonix Corporation

1-Ethyl-1H-imidazole-4,5-dicarbonitrile

  • Cas Number : 133123-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45309.html

Safety Information

MSDS https://synthonix.com/45309.html
Availability details for
Quantity
  • Availability
  • Add to Cart