SY3H3D67D28F Synthonix Corporation

Ethyl 5-bromopyrimidine-2-carboxylate

  • MDL number MFCD12964050
  • Cas Number : 1197193-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45640.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45640.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart