SY3H3D67D29D Synthonix Corporation

Ethyl 4-bromothiophene-2-carboxylate

  • Cas Number : 62224-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45906.html

Safety Information

MSDS https://synthonix.com/45906.html
Availability details for
Quantity
  • Availability
  • Add to Cart