SY3H3D67D2A0 Synthonix Corporation

Ethyl 5-chlorothiophene-2-carboxylate

  • MDL number MFCD00051732
  • Cas Number : 5751-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45915.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45915.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart