SY3H3D67D2AD Synthonix Corporation

Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate

  • MDL number MFCD00010414
  • Cas Number : 64987-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid - Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46048.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46048.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart