SY3H3D67D2B4 Synthonix Corporation

Ethyl 2-chloro-4-methyloxazole-5-carboxylate

  • Cas Number : 78451-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46147.html

Safety Information

MSDS https://synthonix.com/46147.html
Availability details for
Quantity
  • Availability
  • Add to Cart